Learn More
Medchemexpress LLC Alanine dehydrogenase, expressed in E. coli | 9029-06-5 | 1 MG
Supplier: Medchemexpress LLC HYP28541MG
Alanine dehydrogenase, expressed in E. coli, is a microbial enzyme that catalyzes a reversible conversion of L-alanine to pyruvate. It is for research use only. This product is categorized as an endogenous metabolite and a metabolic enzyme/protease.
- Enzyme activity is ≥15 U/mg solid
- One unit corresponds to the amount of enzyme which converts 1 μmol L-alanine per minute at pH 10.0 and 30°C
- Can be stored under recommended conditions as specified in the Certificate of Analysis
- For in vitro use, it is soluble in H2O at 100 mg/mL, requiring ultrasonication