Learn More
Medchemexpress LLC Alanine--tRNA ligase, cytoplasmic (AARS1) | >90.0% | 100 UG

Supplier: Medchemexpress LLC HYP75517100UG
Recombinant full-length human alanyl-tRNA synthetase (AARS1) expressed in Sf9 insect cells with an N-terminal 10xHis tag. Supplied as a filtered solution in 20 mM PB, 500 mM NaCl, pH 7.0, 20% glycerol; purity >90% by reducing SDS-PAGE (sequence M1-N968). Store at -80 °C for long-term storage and avoid repeated freeze-thaw cycles.
- Recombinant full-length human sequence M1-N968.
- Expressed in Sf9 insect cells for eukaryotic processing.
- N-terminal 10xHis tag for affinity purification.
- Purity greater than 90% by reducing SDS-PAGE.
- Formulated in 20 mM PB, 500 mM NaCl, pH 7.0, 20% glycerol, 0.22 μm filtered.
- Store at -80 °C; stable at -20 °C for up to 3 months after opening.
- Suitable for biochemical assays and structural studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.