Learn More
Cayman Chemical AlaproclatehydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 2777010 MG
Encompass_Preferred
An SSRI; inhibits depletion of 5-HT induced by 4-methyl-α-ethyl-m-tyramine in rat cerebral cortex, hippocampus, hypothalamus, and striatum (EC50s = 18, 4, 8, and 12 mg/kg, respectively); inhibits NMDA-evoked currents and depolarization-induced voltage-dependent potassium currents in rat hippocampal neurons (IC50s = 1.1 and 6.9 μM, respectively); increases SIRT1 levels in N2a murine neuroblastoma cells expressing ApoE4 (IC50 = 2.3 μM) and in the hippocampus in the FXFAD-ApoE4 transgenic mouse model of Alzheimer’s disease at 20 mg/kg twice daily; decreases immobility time in the forced swim test in rats at 40 mg/kg
Catalog No. 50-293-9711