Learn More
Apexbio Technology LLC Alarelin Acetate,50mg CAS# 79561-22-1
SDP
Supplier:  Apexbio Technology LLC A315150
Encompass_Preferred
Alarelin (Gonadotrophin-releasing hormone) is a synthetic GnRH agonist. Alarelin is found in higher amounts than that of LH-RH in rat hypophyseal stimulation of gonadotropin secretion in vivo and in vitro and in ovulation inductions. Alarelin is known for its induction of ovulation and is used to treat endmometriosis. Alarelin Acetate is the acetate form of a hypothalamic peptide that stimulates the release of FSH and LH from the pituitary gland. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0246