Learn More
Cayman Chemical Albendazole sulfon-d3 25mg
SDP
Supplier:  Cayman Chemical 3512925 MG
Encompass_Preferred
An internal standard for the quantification of albendazole sulfone by GC- or LC-MS
Catalog No. 50-291-0652