Learn More
MSC Albion Caster Wheel, Cast Iron, 3 in, 1-1/4 in, 3/8 in Axle
Supplier: MSC 00996520
This Albion caster wheel is constructed from cast iron and features a sleeve bearing, designed for reliable performance in demanding environments. With a 3-inch diameter and 1-1/4 inch width, it is suitable for applications requiring durability and high temperature resistance, such as rollers for trays feeding a furnace.
- Made from cast iron for durability
- Sleeve bearing for reliable operation
- 3 inch wheel diameter
- 1-1/4 inch wheel width
- 3/8 inch axle size
- Suitable for temperatures up to 800 F
- Gray wheel color
- 1/2 inch inside diameter