Learn More
MSC Albion Caster Wheel, Forged Steel, 6 x 2-1/2 in, 3/4 in Axle
Supplier: MSC 00996371
This caster wheel is constructed from forged steel, featuring a 6-in diameter and a 2-1/2-in width. It is designed with a 3/4-in axle and a tapered bearing, making it suitable for various industrial applications.
- Tapered bearing for smooth operation.
- 6-in wheel diameter.
- 2-1/2-in wheel width.
- Steel wheel core for durability.
- 3/4-in axle size.
- Forged steel material for high strength.
- Maximum temperature of 800 °F.