Learn More
Alkali Scientific Albumin, Bovine Fraction V [BSA], 5 Grams
Supplier: Alkali Scientific ALB007CP
Bovine Serum Albumin (BSA) Fraction V is a highly purified protein used in a variety of biochemical, molecular biology, and cell culture applications. The 5g format of BSA is ideal for experiments that require a stabilizing agent or carrier protein. BSA is commonly used as a blocking agent in enzyme-linked immunosorbent assays (ELISA), as a stabilizer in protein storage, and as a standard in protein quantification assays. It is also utilized in the preparation of culture media for cell growth, particularly in mammalian cell cultures, where it helps maintain cell viability. Additionally, BSA plays a role in immunological research, such as antibody production and purification.