Learn More
Sigma Aldrich Fine Chemicals Biosciences Bovine Serum Albumin, 9048-46-8, MFCD00130384, 5G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences A94185G/DUP
Encompass
Bovine serum albumin is a model protein in various studies and is made up of 583 amino acids. It is a water-soluble protein with a molecular weight of 66.4kDa. Six α-helices form three homologous domains of BSA. Depending on pH, it undergoes reversible conformational isomerization. The native structure of the protein becomes reactive and flexible on heating. A lyophilized powder, BioReagent, suitable for cell culture
Catalog No. NC0117242