Learn More
Sigma Aldrich Fine Chemicals Biosciences Albumin from human serum l1G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences A43271G
Encompass
Albumin is the most copious protein in blood plasma. Produced in the liver serum albumin serves as a carrier protein for biomolecules like as fatty acids amino acids and steroids. This human serum albumin (HSA) product has been specially tested and evaluated to be low in the following propertiesEssentially protease-free / low protease contentLow alkaline phosphatase contentLow peroxidase contentSerum albumin is used in various applications related to molecular biology notably as an inert protein stabilizer for stock SOLUTIONons of enzymes and proteins of interest. In such applications it is thus important for the HSA to be protease free to maintain the integrity of the protein or enzyme of interest.Protease-free HSA is especially useful for applications likeEnzyme assaysProtein-based assaysProtease-sensitive techniques such as(a) Enzyme immunoassay (EIA)(b) Nucleic acid hybridization(c) Radioimmunoassay (RIA)
Catalog No. 50-290-3847