Learn More
ILS INFRARED LABORATORY SYSTEMS LLC Synermed IR010-R ALBUMIN
Supplier:  ILS INFRARED LABORATORY SYSTEMS LLC IR010R
Encompass_Preferred
Synermed albumin method is a modification of the standard BCG method and is optimized for detection at 660 nm in the near-infrared region of the spectrum. Detection at 660 nm eliminates the effects of hemolysis and minimizes interference from lipemia. Precision and linearity are improved by optimizing the reagent to perform at sample/reagent ratios at which automated devices are most precise.Mono ReagentLiquid-Stable Ready-to-Use ReagentNear-Infrared DetectionHighest PrecisionRoom Temperature StabilitySuperior Dynamic RangeMethod: End Point Reagent BlankWavelength: 600 nmLinearity: 6 mg/dLExpected Values: 3.5-5.0 mg/dL
Catalog No. 22-998-860