Learn More
Apexbio Technology LLC ALC-0159 50mg
SDP
Supplier:  Apexbio Technology LLC B859625
Encompass_Preferred
ALC-0159 is a polyethylene glycol (PEG)-modified lipid compound utilized as a component of lipid nanoparticles (LNP) for mRNA vaccine delivery systems Its primary function is to reduce interactions between lipid particles and plasma proteins through steric hindrance provided by PEG chains thus prolonging particle circulation time in vivo maintaining stability of drug-loaded particles in biological fluids and enhancing the efficiency of mRNA delivery into cells Currently it is widely applied in pharmaceutical formulations and biopharmaceutical research related to nucleic acid vaccines and delivery systems
Catalog No. 50-310-7248