Learn More
Selleck Chemical LLC ALC-0159-E0446-5MG
SDP
Supplier:  Selleck Chemical LLC E04465MG
Encompass_Preferred
ALC-0159 is a polyethylene glycol (PEG) lipid conjugate ALC-0159 allows forming a hydrophilic layer that sterically stabilizes the nanosystem contributing to storage stability and reducing non-specific binding to proteins
Catalog No. 50-313-7042