Learn More
Medchemexpress LLC ALC-0315 | 2036272-55-4 | 99.4% | 766.27 | 1 G
SDP
Supplier:  Medchemexpress LLC HY1381701G
Encompass_Preferred
ALC-0315 is an ionisable aminolipid that facilitates mRNA compaction and aids in its cellular delivery and cytoplasmic release through suspected endosomal destabilization. It can be utilized to form lipid nanoparticle (LNP) delivery vehicles, which have been employed in the research of mRNA COVID-19 vaccines.
  • Ionisable aminolipid
  • Facilitates mRNA compaction
  • Aids mRNA cellular delivery
  • Supports cytoplasmic release via endosomal destabilization
  • Forms lipid nanoparticle delivery vehicles
  • Used in mRNA vaccine research
Catalog No. 50-003-51666