Learn More
Medchemexpress LLC ALC-0315 | 2036272-55-4 | 99.4% | 766.27 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY138170500MG
Encompass_Preferred
ALC-0315 is an ionizable aminolipid that facilitates mRNA compaction, cellular delivery, and cytoplasmic release through endosomal destabilization. It is used to form lipid nanoparticle (LNP) delivery vehicles, which are critical in mRNA vaccine research, including for COVID-19 vaccines.
  • Assists in mRNA cellular delivery
  • Enables mRNA cytoplasmic release
  • Forms lipid nanoparticle (LNP) delivery vehicles
  • Relevant for mRNA vaccine research
Catalog No. 50-003-48598