Learn More
Kyfora Bio ALC-0315 25 MG
Supplier:  Kyfora Bio 1449125
Encompass_Preferred
ALC-0315 is an ionizable lipid designed for the delivery of mRNA in both in vitro and in vivo applications. This lipid is neutral at physiological pH, enabling compatibility with biological systems, but becomes protonated under acidic conditions. Upon protonation, ALC-0315 facilitates electrostatic interactions with nucleic acids and enhances endosomal escape by promoting fusion with endosomal membranes. This property makes it an essential component in lipid nanoparticle (LNP) formulations for effective mRNA delivery. For research use only.Applications: mRNA delivery, LNP formulation
Catalog No. 50-255-9811