Learn More
TARGETMOL CHEMICALS INC ALC-0315 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T927725MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. ALC-0315 is an ionizable amino lipid that has been used in combination with other lipids to form lipid nanoparticles. Lipid-Nanoparticles have been used in the research of mRNA COVID-19 vaccine Purity 99.66%
Catalog No. 50-288-0235