Learn More
Apexbio Technology LLC ALC-0315 50mg
SDP
Supplier:  Apexbio Technology LLC B859525
Encompass_Preferred
ALC-0315 is an ionizable amino lipid used as a component of lipid nanoparticles (LNPs) applied in multiple mRNA vaccine studies Research indicates that ALC-0315 improves nucleic acid drug delivery efficiency and stability by facilitating intracellular transport and cytoplasmic release of mRNA Therefore it shows broad application potential in the delivery of therapeutic mRNA
Catalog No. 50-310-7257