Learn More
Selleck Chemical LLC ALC-0315-E0139-100MG
SDP
Supplier:  Selleck Chemical LLC E0139100MG
Encompass_Preferred
ALC-0315 is an ionisable aminolipid that is responsible for mRNA compaction and aids mRNA cellular delivery and its cytoplasmic release through suspected endosomal destabilization
Catalog No. 50-313-8508