Learn More
Selleck Chemical LLC ALC-0315-E0139-25MG
SDP
Supplier:  Selleck Chemical LLC E013925MG
Encompass_Preferred
ALC-0315 is an ionisable aminolipid that is responsible for mRNA compaction and aids mRNA cellular delivery and its cytoplasmic release through suspected endosomal destabilization
Catalog No. 50-313-6836