Learn More
Medchemexpress LLC Alclofenac | 22131-79-9 | 99.4% | 226.66 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYW009706
Encompass_Preferred
Alclofenac (Allopydin) is a non-steroidal anti-inflammatory agent (NSAID). It acts as a prostaglandin H2 synthase inhibitor. It is used in research for rheumatoid arthritis, degenerative joint disease, and ankylosing spondylitis.
  • Non-steroidal anti-inflammatory agent (NSAID)
  • Prostaglandin H2 synthase inhibitor
  • Used in research for rheumatoid arthritis, degenerative joint disease, and ankylosing spondylitis
Catalog No. 50-004-53261