Learn More
Volu Sol Alcoholic Giemsa Stain, 16 oz / 500 mL, Volu-Sol™
SDP
Supplier:  Volu Sol VAG016
Encompass_Preferred
Giemsa Stain, modified.  Blood films staining; the nucleus and cytoplasm of white blood cells take on the characteristic blue or pink coloration
Catalog No. 50-420-787