Learn More
Medchemexpress LLC Ald-Ph-amido-C2-PEG3-azide | 1807540-88-0 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY130667
Encompass_Preferred
Ald-Ph-amido-C2-PEG3-azide is a laboratory chemical used for the manufacture of substances and is for research use only. It is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. This click chemistry reagent contains an azide group, enabling copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with alkyne-containing molecules, and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups.
  • Used in the synthesis of PROTACs.
  • Functions as a PEG-based PROTAC linker.
  • Contains an azide group for click chemistry reactions.
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc).
  • Facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups.
  • Exploits the intracellular ubiquitin-proteasome system to degrade target proteins.
  • For research use only.
Catalog No. 50-003-41661