Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ald-Ph-amido-C2-PEG3-azide | 1807540-88-0 | MFCD28155209 | 98.9% | 350.37 g/mol | C16H22N4O5 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY1306675MG

Encompass_Preferred

Ald-Ph-amido-C2-PEG3-azide is a PEG-based PROTAC linker and click chemistry reagent bearing an azide functional group. It enables bioconjugation and PROTAC assembly via copper-catalyzed azide-alkyne cycloaddition (CuAAC) or strain-promoted azide-alkyne cycloaddition (SPAAC), and provides a soluble, flexible PEG spacer for linker construction.

  • Contains an azide functional group for click chemistry.
  • Compatible with CuAAC and SPAAC conjugation methods.
  • PEG3 spacer improves solubility and linker flexibility.
  • High purity suitable for research applications (>98%).
  • Available in small milligram pack sizes for discovery work.
  • Molecular weight 350.37 g/mol; formula C16H22N4O5.

Catalog No. 50-002-62776


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.