Learn More
Medchemexpress LLC HY-130107 50mg Medchemexpress, Ald-Ph-NHS ester CAS:60444-78-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13010750MG
Encompass_Preferred
Medchemexpress, HY-130107 50mg Ald-Ph-NHS ester CAS:60444-78-2 Ald-Ph-NHS ester is a nonclaevable linker for antibody-drug-conjugation (ADC). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1224