Learn More
Medchemexpress LLC Ald-Ph-PEG4-Boc | 1807518-64-4 | >98.0% | C23H35NO8 | 500 MG
Supplier: Medchemexpress LLC HY140626500MG
Ald-Ph-PEG4-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. This product is for research use only and not sold to patients.
- PEG-based PROTAC linker
- Used in the synthesis of PROTACs
- Exploits the intracellular ubiquitin-proteasome system
- Selectively degrades target proteins
- For research use only
- Not for sale in certain territories due to controlled substance status