Learn More
Medchemexpress LLC Aldastotug 25mg | 25mg
SDP
Supplier:  Medchemexpress LLC HYP99136425MG
Encompass_Preferred
Aldastotug (PYX-106 BSI-060T) is a human monoclonal antibody (mAb) targeting Siglec-15/CD33L3 Aldastotug reverses Siglec-15-mediated immunosuppression and enhances T-cell proliferation Aldastotug induces significantly increased levels of IFN and TNF in T cells Aldastotug can be used in cancer immunity research Recommended isotype control Human IgG1 kappa Isotype Control (HY-P99001)[1]
Catalog No. 50-003-23184