Learn More
Medchemexpress LLC Aldehyde-benzyl-peg5-alkyne | 1378928-83-6 | 97.1% | C18H24O6 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13614010MG
Encompass_Preferred
Aldehyde-benzyl-PEG5-alkyne is a PEG-based bifunctional linker bearing an aromatic aldehyde and a terminal alkyne, designed for orthogonal conjugation strategies. The aldehyde enables formation of oximes or Schiff bases, while the alkyne participates in copper-catalyzed azide-alkyne cycloaddition (CuAAC). The reagent is supplied as a purified solid for use in PROTAC synthesis, bioconjugation, and click chemistry workflows.
  • Contains aldehyde and terminal alkyne functional groups for orthogonal conjugation
  • PEG5 spacer improves solubility and reduces steric hindrance
  • Enables CuAAC click chemistry and aldehyde-based ligation
  • High purity suitable for research and synthetic applications
  • Available in small-mass quantities for discovery chemistry
Catalog No. 50-002-59901