Learn More
Sigma Aldrich Fine Chemicals Biosciences Aldehyde Dehydrogenase fro
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 10171832001
Encompass
Aldehyde dehydrogenase (ALDH) is a soluble enzyme and its activity depends on potassium ions and cysteine. The family of ALDH comprises of 19 genes in human. ALDH gene is expressed in the nucleus cytosol mitochondria and endoplasmic reticulum of the cell. ALDH is a component of nicotinamide adenine dinucleotide (NADH) and nicotinamide adenine dinucleotide phosphate (NADPH) recycling systems.
Catalog No. 50-290-1923