Learn More
Cayman Chemical Aldehyde Reactive Probe (trifluoroacetate salt), 627090-10-2, 5 mg
SDP
Supplier:  Cayman Chemical 100093505
Encompass_Preferred
Molecular Formula C12H21N5O4S • CF3COOH, Purity ≥95%, Formula Weight 445.4, Synonyms: (6aR)-hexahydro-2-oxo-2-[(aminooxy)acetyl]hydrazide,1H-thieno[3aS,4S-d]imidazole-4-pentanoic acid, trifluoroacetate salt, ARP, O-(Biotinylcarbazoylmethyl) Hydroxylamine; A biotinylated reagent used for the detection and quantification of apurinic/apyrimidinic (AP) sites in damaged DNA; reacts with aldehyde groups formed when reactive oxygen species depurinate DNA, thereby covalently linking biotin to these AP sites.
Catalog No. NC0164582