Learn More
Cayman Chemical ALDH Enzyme MIxture 400UG
SDP
Supplier:  Cayman Chemical 700803400 UG
Encompass_Preferred
An ionizable cationic lipid (pKa 6.4) has been used in the generation of LNPs for the delivery of mRNA in vitro and in vivo LNPs containing E12CA1A3 and encapsulating an mRNA reporter induce luciferase reporter expression in mouse BMDCs and mice LNPs containing E12CA1A3 are cleared more rapidly from the liver than LNPs containing DLin-MC3-DMA in mice
Catalog No. 50-289-2775