Learn More
ABclonal Technology ALDH1A1 Rabbit mAb
SDP
Supplier:  ABclonal Technology A2235120UL
Encompass_Preferred
The protein encoded by this gene belongs to the aldehyde dehydrogenase family Aldehyde dehydrogenase is the next enzyme after alcohol dehydrogenase in the major pathway of alcohol metabolism There are two major aldehyde dehydrogenase isozymes in the liver cytosolic and mitochondrial which are encoded by distinct genes and can be distinguished by their electrophoretic mobility kinetic properties and subcellular localization This gene encodes the cytosolic isozyme Studies in mice show that through its role in retinol metabolism this gene may also be involved in the regulation of the metabolic responses to high-fat diet
Catalog No. 50-000-05898