Learn More
TARGETMOL CHEMICALS INC ALDH1A3-IN-2 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T6024010MG
Encompass_Preferred
Also available in 5 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. ALDH1A3-IN-2 is a potent ALDH1A3 inhibitor with an IC50 of 0.30 uM.ALDH1A3-IN-2 has potential for cancer disease research. purity: 99%
Catalog No. 50-257-6625