Learn More
Medchemexpress LLC ALDH1A3-IN-3 | 18962-05-5 | MFCD00052357 | 99.3% | 164.20 g/mol | C10H12O2 | 25 G

Supplier: Medchemexpress LLC 25GHYW017186
ALDH1A3-IN-3 is a small-molecule research inhibitor of aldehyde dehydrogenase 1A3 with reported inhibitory activity (IC50 = 0.26 μM). It is used in biochemical and cellular studies related to aldehyde dehydrogenase biology and has documented solubility and storage recommendations for both in vitro and in vivo work.
- Potent inhibitor of ALDH1A3 (IC50 = 0.26 μM).
- Also a substrate for ALDH3A1, which may limit selectivity in some assays.
- Molecular formula C10H12O2 and molecular weight 164.20 g/mol.
- High reported purity (99.26%).
- Soluble in DMSO (100 mg/mL); documented in vivo formulations available.
- Store at 4°C under nitrogen; in-solvent storage recommendations provided.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.