Learn More
Medchemexpress LLC Aldh1a3-in-3 (compound 16) | 18962-05-5 | MFCD00052357 | 99.3% | 164.20 | C10H12O2 | 10 G

Supplier: Medchemexpress LLC HYW01718610G
ALDH1A3-IN-3 is a small-molecule inhibitor of aldehyde dehydrogenase 1A3 used in biochemical and cellular research, notably in prostate cancer studies. It inhibits ALDH1A3 with an IC50 of 0.26 μM but is also a substrate of ALDH3A1, which limits its selectivity in some applications.
- Potent ALDH1A3 inhibitor (IC50 0.26 μM).
- Substrate of ALDH3A1, limiting selectivity.
- High purity (99.3%).
- Molecular weight 164.20; formula C10H12O2.
- Available in multiple pack sizes including 10 g.
- Storage: store at 4°C under nitrogen; in solution, store at -80°C (6 months) or -20°C (1 month).
- For research use only; not for human use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.