Learn More
Cayman Chemical AldIcarb SulfoxIde 50mg
SDP
Supplier:  Cayman Chemical 2421150 MG
Encompass_Preferred
A metabolite and degradation product of aldicarb; reduces the number of male T. semipenetrans second-stage larvae that reach the third, fourth, and adult stages of development at 10 μg/ml; inhibits carboxylesterase and cholinesterase in zebrafish (IC50 = 10 μM for both)
Catalog No. 50-293-6117