Learn More
Medchemexpress LLC Aldometanib | 2904601-67-6 | 99.7% | 593.45 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY14818910MM/1ML
Encompass_Preferred
Aldometanib (LXY-05-029) is an orally active aldolase inhibitor that activates lysosomal adenosine monophosphate-activated protein kinase (AMPK) and lowers blood glucose levels. It can be used for the research of metabolic homeostasis, alleviating fatty liver and nonalcoholic steatohepatitis, and extending lifespan in C. elegans.
  • Activates lysosomal AMPK
  • Lowers blood glucose levels
  • Suitable for research on metabolic homeostasis
  • Alleviates fatty liver and nonalcoholic steatohepatitis
  • Extends lifespan in C. elegans
  • Reduces blood glucose in lean and obese hyperglycemic mice
  • Alleviates liver fibrosis in NASH mice
Catalog No. 50-003-63711