Learn More
Enzo Life Sciences Aldosterone ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI900173
Encompass_Preferred
The Aldosterone EIA kit is a colorimetric competitive enzyme immunoassay kit with results overnight + 1 hour. Sensitivity: 4.7 pg/ml (range 3.9 - 250 pg/ml). Assay Time: Overnight + 1 hour. Applications: ELISA, Colorimetric detection. Species Reactivity: Species independent. Contents: DxS IgG Microtiter plate, Conjugate, Antibody, Assay buffer concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution
Catalog No. 50-946-889