Learn More
Medchemexpress LLC HY-16261 10mg , Aldoxorubicin CAS:1361644-26-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1626110MG
Encompass_Preferred
HY-16261 10mg Aldoxorubicin CAS: 1361644-26-9 Aldoxorubicin (INNO-206) is an albumin-binding prodrug of Doxorubicin (DNA topoisomerase II inhibitor), which is released from albumin under acidic conditions. Aldoxorubicin (INNO-206) has potent antitumor activities in various cancer cell lines and in murine tumor models. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9066