Learn More
Sigma Aldrich Fine Chemicals Biosciences AldrichR BenchclampTM 3-pr1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z6898311EA
Encompass
Made entirely of electropolished stainless steel for excellent chemical resistance and strength. Round extension arms allow the clamps to be rotated 360. Extension arms allow placement of apparatus at various distances from lab frames. Supplied with vinyl and fiberglass prong covers for high temperature applications.NOTE An additional clamp holder is required to attach clamps to frames or other apparatus.
Catalog No. 50-290-1397