Learn More
Sigma Aldrich Fine Chemicals Biosciences AldrichR check-valve bubbl1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z5518641EA
Encompass
Check-valve ball prevents oil from being drawn into the system. Order replacement SafetyBarb hose connectors Z547786 (straight) or Z547883 (angled).
Catalog No. 50-290-5998