Learn More
Medchemexpress LLC HY-13011A 50mg , Alectinib (Hydrochloride) CAS:1256589-74-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13011A50MG
Encompass_Preferred
HY-13011A 50mg Alectinib (Hydrochloride) CAS: 1256589-74-8 Alectinib Hydrochloride (CH5424802 Hydrochloride; RO5424802 Hydrochloride; AF-802 Hydrochloride) is a potent, selective, and orally available ALK inhibitor with an IC50 of 1.9 nM and a Kd value of 2.4 nM (in an ATP-competitive manner), and also inhibits ALK F1174L and ALK R1275Q with IC50s of 1 nM and 3.5 nM, respectively. Alectinib demonstrates effective central nervous system (CNS) penetration. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0031