Learn More
Apexbio Technology LLC Alectinib Hydrochloride 1256589-74-8 25mg
SDP
Supplier:  Apexbio Technology LLC A464925
Encompass_Preferred
Alectinib Hydrochloride (CAS 1256589-74-8) is an orally administered selective inhibitor of anaplastic lymphoma kinase (ALK) that acts by competitively binding to the ATP-binding site of the ALK kinase domain thereby disrupting downstream signaling pathways In vitro studies demonstrate potent inhibition of ALK with an IC50 of approximately 1 9 nM and it also effectively targets ALK mutant variants F1174L (IC50 1 nM) and R1275Q (IC50 3 5 nM) Owing to its ability to penetrate the central nervous system alectinib hydrochloride is utilized in cancer research to investigate ALK-dependent tumorigenesis and mechanisms of drug resistance
Catalog No. 50-000-94943