Learn More
Enzo Life Sciences Alectinib. hydrochloride (25mg). CAS: 1256589-74-8
SDP
Supplier:  Enzo Life Sciences ENZCHM2130025
Encompass_Preferred
Alectinib hydrochloride is a selective anaplastic lymphoma kinase (ALK) inhibitor leading to inhibition of tumor cell proliferation and induction of cell death.  Purity: ≥99% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-3537