Learn More
TARGETMOL CHEMICALS INC ALEGLITAZAR 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T141765MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Aleglitazar (R1439) (R1439) is a potent dual PPAR(alpha)/(gamma) agonist with IC50s of 38 nM and 19 nM for human PPARa and PPAR(gamma) respectively. Aleglitazar can be used for the research of type II diabetes. purity: 99%
Catalog No. 50-258-1308