Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Alendronate sodium hydrate | 121268-17-5 | 99.6% | 325.12 | 1 ML
SDP

Supplier:  Medchemexpress LLC HY1110110MM/1ML

Encompass_Preferred

Alendronate sodium hydrate is a farnesyl diphosphate synthase inhibitor with an IC50 of 460 nM. It directly acts on osteoclasts, inhibiting a rate-limiting step in the cholesterol biosynthesis pathway, which is crucial for osteoclast function. It inhibits the isoprenoid biosynthesis pathway and interferes with protein prenylation by reducing geranylgeranyl diphosphate levels. Alendronate can cause erosions in the rabbit stomach but not antral ulceration in rats.

  • Solid appearance, white to off-white color
  • Chemical formula: C4H18NNaO10P2
  • Soluble in H2O: ≥ 28.57 mg/mL (87.88 mM)
  • Shipping at room temperature in continental US; may vary elsewhere
  • Storage at 4°C, sealed, away from moisture
  • In solvent storage at -80°C for 6 months or -20°C for 1 month (sealed, away from moisture)

Catalog No. 50-003-43668


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.