Learn More
Medchemexpress LLC Alendronic acid | 66376-36-1 | 98.0% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0631100MG
Encompass_Preferred
Alendronic acid is an orally active bisphosphonate that binds to bone surfaces and inhibits bone resorption by osteoclasts. It induces skeletal alterations in the chicken embryonic development model and can be used for osteoporosis research.
  • Inhibits bone resorption by osteoclasts.
  • Induces skeletal alterations in chicken embryonic development model.
  • Useful for osteoporosis research.
  • Orally active bisphosphonate.
Catalog No. 50-004-08181