Learn More
Selleck Chemical LLC Alendronic Acid
SDP
Supplier:  Selleck Chemical LLC S55361G
Encompass_Preferred
Alendronic acid (alendronate) is a nitrogen-containing second generation bisphosphonate used for osteoporosis osteogenesis imperfecta and several other bone diseases
Catalog No. 50-000-03807