Learn More
Medchemexpress LLC Aleuritic acid | 533-87-9 | 25 MG

Supplier: Medchemexpress LLC 25MGHYN7076
Aleuritic acid (C16H32O5, MW: 304.42, CAS: 533-87-9) is a white to off-white solid that serves as a major ingredient in Croton crassifolius Geisel. and is widely utilized in the perfumery industry. This compound is intended for research use only and not for medical applications.
- High purity, 99.89%
- Molecular formula: C16H32O5
- Molecular weight: 304.42
- Appearance: White to off-white solid
- Melting point: 100-101 °C
- Soluble in DMSO (100 mg/mL)
- Powder storage: -20°C for 3 years, 4°C for 2 years
- Solution storage: -80°C for 6 months, -20°C for 1 month
- For laboratory chemical use
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.