Learn More
Biolegend Alexa Fluor 488 anti-GFAP
Supplier:  Biolegend 837508
Encompass_Preferred
Alexa Fluor 488 anti-GFAP [SMI 25]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Mouse, Rat; Apps: IHC-P; Size: 100 ug
Catalog No. 50-207-4407